6-Iodoquinazoline ≥95%

Supplier: Matrix Scientific

072240-250MG 072240-1G 072240-5G
102523-840EA 569.23 USD
102523-840 102523-842 102523-844
6-Iodoquinazoline ≥95%
6-Iodoquinazoline

Hazard: Irritant

Formula: C₈H₅IN₂
MW: 256.0400 g/mol
MDL Number: MFCD09834646
CAS Number: 848841-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR