4-(4-Bromopyrimidin-2-yl)morpholine ≥95%

Supplier: Matrix Scientific

072239-250MG 072239-1G 072239-5G
102523-834EA 400 USD
102523-834 102523-836 102523-838
4-(4-Bromopyrimidin-2-yl)morpholine ≥95%
4-(4-Bromopyrimidin-2-yl)morpholine

Hazard: Irritant

Formula: C₈H₁₀BrN₃O
MDL Number: MFCD03861153
CAS Number: 663194-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR