2-Morpholinopyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

072238-250MG 072238-1G 072238-5G
102523-828EA 427.69 USD
102523-828 102523-830 102523-832
2-Morpholinopyrimidin-4-ol ≥95%
2-Morpholinopyrimidin-4-ol

Hazard: Irritant

Formula: C₈H₁₁N₃O₂
MW: 181.1900 g/mol
MDL Number: MFCD00234443
CAS Number: 19810-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR