6-Phenyl-5H-pyrrolo[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

072236-250MG 072236-1G
102523-818EA 684.62 USD
102523-818 102523-820
6-Phenyl-5H-pyrrolo[2,3-b]pyrazine ≥95%
6-Phenyl-5H-pyrrolo[2,3-b]pyrazine

Hazard: Irritant

Formula: C₁₂H₉N₃
CAS Number: 78605-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR