4-Amino-5-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072234-1G 072234-5G
102523-808EA 160 USD
102523-808 102523-810
4-Amino-5-bromopyrimidine ≥95%
4-Amino-5-bromopyrimidine

Hazard: Irritant

Formula: C₄H₄BrN₃
MW: 174.00 g/mol
MDL Number: MFCD00233962
CAS Number: 1439-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR