2-Amino-5-bromo-3-(diethylamino)pyrazine ≥95%

Supplier: Matrix Scientific

072233-1G 072233-5G 072233-10G
102523-802EA 355.38 USD
102523-802 102523-804 102523-806
2-Amino-5-bromo-3-(diethylamino)pyrazine ≥95%
2-Amino-5-bromo-3-(diethylamino)pyrazine

Hazard: Irritant

Formula: C₈H₁₃BrN₄
CAS Number: 912773-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR