6-Bromo-N2,N2-dimethylpyrazine-2,3-diamine ≥95%
Supplier: Matrix Scientific
072232-1G
072232-5G
072232-10G
102523-796EA
355.38
USD
102523-796
102523-798
102523-800
6-Bromo-N2,N2-dimethylpyrazine-2,3-diamine ≥95%
6-Bromo-N2,N2-dimethylpyrazine-2,3-diamine
Hazard: Irritant
Formula:
C₆H₉BrN₄ |
CAS Number:
89641-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...