2-Amino-5-bromo-3-(ethylamino)pyrazine ≥95%

Supplier: Matrix Scientific

072231-1G 072231-5G 072231-10G
102523-790EA 355.38 USD
102523-790 102523-792 102523-794
2-Amino-5-bromo-3-(ethylamino)pyrazine ≥95%
2-Amino-5-bromo-3-(ethylamino)pyrazine

Hazard: Irritant

Formula: C₆H₉BrN₄
CAS Number: 117719-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR