3,5-Dibromo-2-hydroxypyrazine ≥95%

Supplier: Matrix Scientific

072226-1G 072226-5G
102523-762EA 30.77 USD
102523-762 102523-764
3,5-Dibromo-2-hydroxypyrazine ≥95%
3,5-Dibromo-2-hydroxypyrazine

Hazard: Irritant/Light Sensitive

Formula: C4H2Br2N2O
MW: 253.8800 g/mol
Melting Pt: 174…178 °C
MDL Number: MFCD07371406
CAS Number: 21943-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR