2,3-Diamino-5-bromopyrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromopyrazine-2,3-diamine
072225-1G
072225-5G
072225-10G
102523-756EA
427.69
USD
102523-756
102523-758
102523-760
2,3-Diamino-5-bromopyrazine ≥95%
2,3-Diamino-5-bromopyrazine
Hazard: Irritant
Formula:
C₄H₅BrN₄ MW: 189.0100 g/mol |
MDL Number:
MFCD06245575 CAS Number: 89123-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...