2-Amino-3-(dimethylamino)pyrazine ≥95%

Supplier: Matrix Scientific

072223-250MG 072223-1G 072223-5G
102523-744EA 355.38 USD
102523-744 102523-746 102523-748
2-Amino-3-(dimethylamino)pyrazine ≥95%
2-Amino-3-(dimethylamino)pyrazine

Hazard: Irritant

Formula: C₆H₁₀N₄
CAS Number: 89488-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR