2-Amino-3-(ethylamino)pyrazine ≥95%

Supplier: Matrix Scientific

072222-250MG 072222-1G 072222-5G
102523-738EA 355.38 USD
102523-738 102523-740 102523-742
2-Amino-3-(ethylamino)pyrazine ≥95%
2-Amino-3-(ethylamino)pyrazine

Hazard: Irritant

Formula: C₆H₁₀N₄
MDL Number: MFCD08460071
CAS Number: 912773-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR