2-Amino-3-methoxypyrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methoxypyrazin-2-amine
072218-1G
072218-5G
102523-716EA
123.08
USD
102523-716
102523-718
2-Amino-3-methoxypyrazine ≥95%
2-Amino-3-methoxypyrazine
Hazard: Irritant
Formula:
C₅H₇N₃O MW: 125.1300 g/mol |
MDL Number:
MFCD05722793 CAS Number: 4774-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...