2,3-Diaminopyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrazine-2,3-diamine

072217-1G 072217-5G
102523-712EA 152.31 USD
102523-712 102523-714
2,3-Diaminopyrazine ≥95%
2,3-Diaminopyrazine

Hazard: Irritant

Formula: C₄H₆N₄
MW: 110.1172 g/mol
MDL Number: MFCD07778385
CAS Number: 13134-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR