Pyrimidine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Formylpyrimidine, 4-Formyl-1,3-diazine, 4-Pyrimidinecarboxaldehyde

072216-250MG 072216-1G 072216-5G
102523-704EA 212.31 USD
102523-704 102523-706 102523-708
Pyrimidine-4-carbaldehyde ≥95%
Pyrimidine-4-carbaldehyde

Hazard: Irritant

Formula: C₅H₄N₂O
MW: 108.1 g/mol
MDL Number: MFCD03789624
CAS Number: 2435-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR