N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-pyridyl)-2-pyrimidineamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine, N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine

072213-1G 072213-5G
102523-688EA 123.08 USD
102523-688 102523-690
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥95%
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine

Hazard: Irritant

Formula: C₁₆H₁₅N₅
MDL Number: MFCD09028125
CAS Number: 152460-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR