N-(4-Oxocyclohexyl)acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Acetamidocyclohexanone

072212-5G 072212-10G 072212-100G
102523-682EA 90.77 USD
102523-682 102523-684 102523-686
N-(4-Oxocyclohexyl)acetamide ≥95%
N-(4-Oxocyclohexyl)acetamide

Hazard: Irritant

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
MDL Number: MFCD03703462
CAS Number: 27514-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR