tert-Butyl methyl(4-oxocyclohexyl)carbamate ≥95%

Supplier: Matrix Scientific

072211-1G 072211-5G
102523-678EA 150.77 USD
102523-678 102523-680
tert-Butyl methyl(4-oxocyclohexyl)carbamate ≥95%
tert-Butyl methyl(4-oxocyclohexyl)carbamate

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MW: 227.3000 g/mol
MDL Number: MFCD09037870
CAS Number: 400899-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR