Methyl-3-chloropicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-chloropicolinate, Methyl 3-chloropyridine-2-carboxylate

072210-250MG 072210-1G 072210-5G
102523-672EA 498.46 USD
102523-672 102523-674 102523-676
Methyl-3-chloropicolinate ≥95%
Methyl 3-chloropicolinate

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.5810 g/mol
MDL Number: MFCD09750958
CAS Number: 116383-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR