(S)-N-3-Cyanophenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: H-Phe(3-CN)-OH

072207-1G 072207-5G 072207-10G
102523-654EA 284.62 USD
102523-654 102523-656 102523-658
(S)-N-3-Cyanophenylalanine ≥95%
(S)-N-3-Cyanophenylalanine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
MDL Number: MFCD01860885
CAS Number: 57213-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR