Ethyl-6-chloro-4-methylpyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

072204-1G 072204-5G
102523-638EA 276.92 USD
102523-638 102523-640
Ethyl-6-chloro-4-methylpyridine-2-carboxylate ≥95%
Ethyl-6-chloro-4-methylpyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₀ClNO₂
CAS Number: 1122090-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR