Ethyl-5-amino-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-amino-1H-pyrazole-4-carboxylate

072203-1G 072203-5G 072203-10G
102523-630EA 364.62 USD
102523-630 102523-632 102523-634
Ethyl-5-amino-1H-pyrazole-4-carboxylate ≥95%
Ethyl-5-amino-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₉N₃O₂
MW: 155.1546 g/mol
MDL Number: MFCD00114562
CAS Number: 1260243-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR