Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-methyl-5-nitro-3-pyridinecarboxylate

072202-250MG 072202-1G 072202-5G
102523-624EA 355.38 USD
102523-624 102523-626 102523-628
Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate ≥95%
Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₁₀N₂O₄
MW: 210.1867 g/mol
MDL Number: MFCD00753850
CAS Number: 51984-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR