Ethyl-2-bromo-6-benzothiazolecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromo-6-benzothiazolecarboxylate

072201-1G 072201-5G 072201-10G
102523-618EA 640 USD
102523-618 102523-620 102523-622
Ethyl-2-bromo-6-benzothiazolecarboxylate ≥95%
Ethyl-2-bromo-6-benzothiazolecarboxylate

Hazard: Irritant

Formula: C₁₀H₈BrNO₂S
MW: 286.1500 g/mol
MDL Number: MFCD08669850
CAS Number: 99073-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR