Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate
072200-1G
072200-5G
072200-10G
102523-612EA
249.23
USD
102523-612
102523-614
102523-616
Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥95%
Ethyl 2,4-dimethylpyrrole-3-carboxylate
Hazard: Irritant
Formula:
C₉H₁₃NO₂ |
MDL Number:
MFCD01365811 CAS Number: 2199-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...