D-β-Homoserine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Amino-4-hydroxybutanoic acid

072199-1G 072199-5G 072199-10G
102523-606EA 307.69 USD
102523-606 102523-608 102523-610
D-β-Homoserine ≥95%
D-beta-Homoserine

Hazard: Irritant

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00270242
CAS Number: 16504-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR