6-Phenylpicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Phenylpicolinic acid, 6-Phenylpyridine-2-carboxylic acid

072196-1G 072196-5G
102523-594EA 533.85 USD
102523-594 102523-596
6-Phenylpicolinic acid ≥95%
6-Phenylpicolinic acid

Hazard: Irritant

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD04114206
CAS Number: 39774-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR