6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one ≥95%

Supplier: Matrix Scientific

072195-250MG 072195-1G
102523-590EA 364.62 USD
102523-590 102523-592
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one ≥95%
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2000 g/mol
MDL Number: MFCD04114865
CAS Number: 22246-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR