6-Hydroxybenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Benzothiazolol

072194-1G 072194-5G
102523-584EA 46.15 USD
102523-584 102523-586
6-Hydroxybenzothiazole ≥95%
6-Hydroxybenzothiazole

Hazard: Irritant

Formula: C₇H₅NOS
MW: 151.1857 g/mol
MDL Number: MFCD00057883
CAS Number: 13599-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR