6-Chloro-4-methylpyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072193-250MG 072193-1G 072193-5G
102523-578EA 423.69 USD
102523-578 102523-580 102523-582
6-Chloro-4-methylpyridine-2-carboxylic acid ≥95%
6-Chloro-4-methylpyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD09834827
CAS Number: 324028-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR