1-Amino-6-bromoisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromoisoquinolin-1-ylamine

072191-250MG 072191-1G
102523-568EA 184.62 USD
102523-568 102523-570
1-Amino-6-bromoisoquinoline ≥95%
1-Amino-6-bromoisoquinoline

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.0700 g/mol
MDL Number: MFCD07374395
CAS Number: 215453-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR