6-Bromoimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

072190-1G 072190-5G 072190-10G
102523-562EA 692.31 USD
102523-562 102523-564 102523-566
6-Bromoimidazo[1,2-a]pyrazine ≥95%
6-Bromoimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD09750036
CAS Number: 912773-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR