6-Bromobenzo[d]isoxazol-3-amine ≥95%

Supplier: Matrix Scientific

072188-1G 072188-5G
102523-552EA 640 USD
102523-552 102523-554
6-Bromobenzo[d]isoxazol-3-amine ≥95%
6-Bromobenzo[d]isoxazol-3-amine

Hazard: Irritant

Formula: C₇H₅BrN₂O
MW: 213.0314 g/mol
MDL Number: MFCD09788587
CAS Number: 177995-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR