2-Amino-6-bromoquinazoline ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Bromoquinazolin-2-amine
072186-1G
072186-5G
102523-542EA
173.85
USD
102523-542
102523-544
2-Amino-6-bromoquinazoline ≥95%
2-Amino-6-bromoquinazoline
Hazard: Irritant
Formula:
C₈H₆BrN₃ MW: 224.0600 g/mol |
MDL Number:
MFCD04114089 CAS Number: 190273-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...