6-Aminobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

072185-1G 072185-5G
102523-536EA 44.62 USD
102523-536 102523-538
6-Aminobenzothiazole ≥95%
6-Aminobenzothiazole

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00015461
CAS Number: 533-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR