5-Iodo-o-toluic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-lodo-2-methylbenzoic acid, 5-Iodo-2-methylbenzoic acid
072181-5G
072181-25G
072181-100G
102523-516EA
23.08
USD
102523-516
102663-868
102663-870
5-Iodo-o-toluic acid ≥95%
5-Iodo-o-toluic acid
Hazard: Irritant
Formula:
C₈H₇IO₂ MW: 262.0400 g/mol |
MDL Number:
MFCD01570258 CAS Number: 54811-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...