5-Hydroxy-2-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-nitropyridin-3-ol

072180-250MG 072180-1G 072180-5G
102523-508EA 358.46 USD
102523-508 102523-510 102523-512
5-Hydroxy-2-nitropyridine ≥95%
5-Hydroxy-2-nitropyridine

Hazard: Irritant

Formula: C5H4N2O3
MW: 140.0969 g/mol
MDL Number: MFCD03032441
CAS Number: 15206-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR