5-Fluoro-4-hydroxypyrimidine ≥95%

Supplier: Matrix Scientific

072179-1G 072179-5G 072179-10G
102523-502EA 90.77 USD
102523-502 102523-504 102523-506
5-Fluoro-4-hydroxypyrimidine ≥95%
5-Fluoro-4-hydroxypyrimidine

Hazard: Irritant

Formula: C4H3FN2O
MW: 114.778 g/mol
MDL Number: MFCD00223674
CAS Number: 671-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR