5-Fluoro-3-iodo-1H-indazole ≥95%
Supplier: Matrix Scientific
072178-1G
072178-5G
102523-498EA
463.08
USD
102523-498
102523-500
5-Fluoro-3-iodo-1H-indazole ≥95%
5-Fluoro-3-iodo-1H-indazole
Hazard: Irritant
Formula:
C₇H₄FIN₂ MW: 262.02 g/mol |
MDL Number:
MFCD06739145 CAS Number: 858629-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...