5-Bromobenzo[d]isoxazol-3-ylamine ≥95%

Supplier: Matrix Scientific

072177-1G 072177-5G
102523-494EA 96.92 USD
102523-494 102523-496
5-Bromobenzo[d]isoxazol-3-ylamine ≥95%
5-Bromobenzo[d]isoxazol-3-ylamine

Hazard: Irritant

Formula: C₇H₅BrN₂O
MW: 213.0300 g/mol
MDL Number: MFCD09834823
CAS Number: 455280-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR