5-Bromo-4-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

072176-250MG 072176-1G 072176-5G
102523-488EA 498.46 USD
102523-488 102523-490 102523-492
5-Bromo-4-pyrimidinecarboxylic acid ≥95%
5-Bromo-4-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₅H₃BrN₂O₂
MDL Number: MFCD09754104
CAS Number: 64224-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR