5-Amino-6-methyl-2-benzimidazolone ≥95%

Supplier: Matrix Scientific

072175-1G 072175-5G 072175-10G
102523-482EA 278.46 USD
102523-482 102523-484 102523-486
5-Amino-6-methyl-2-benzimidazolone ≥95%
5-Amino-6-methyl-2-benzimidazolone

Hazard: Irritant

Formula: C₈H₇N₃O
MDL Number: MFCD09996978
CAS Number: 67014-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR