5,7-Dihydroxyimidazo[1,2-a]pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyrimidine-5,7-diol

072174-250MG 072174-1G 072174-5G
102523-476EA 166.15 USD
102523-476 102523-478 102523-480
5,7-Dihydroxyimidazo[1,2-a]pyrimidine ≥95%
5,7-Dihydroxyimidazo[1,2-a]pyrimidine

Hazard: Irritant

Formula: C₆H₅N₃O₂
MW: 151.13 g/mol
MDL Number: MFCD09834820
CAS Number: 51647-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR