4-Phenyl-2-pyrrolidone ≥95%

Supplier: Matrix Scientific

072172-1G 072172-5G
102523-466EA 183.08 USD
102523-466 102523-468
4-Phenyl-2-pyrrolidone ≥95%
4-Phenyl-2-pyrrolidone

Hazard: Irritant

Formula: C₁₀H₁₁NO
MDL Number: MFCD01687226
CAS Number: 1198-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR