1H-Pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

072168-1G 072168-5G
102523-450EA 103.08 USD
102523-450 102523-452
1H-Pyrazole-4-carbonitrile ≥95%
1H-Pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₄H₃N₃
MW: 93.0900 g/mol
MDL Number: MFCD00040250
CAS Number: 31108-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR