4-Chloro-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

072166-1G 072166-5G
102523-440EA 60 USD
102523-440 102523-442
4-Chloro-6-methylpyrimidine ≥95%
4-Chloro-6-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅ClN₂
MDL Number: MFCD02322991
CAS Number: 3435-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR