4-Chloro-3-nitroquinoline ≥95%

Supplier: Matrix Scientific

072162-1G 072162-5G
102523-422EA 73.85 USD
102523-422 102523-424
4-Chloro-3-nitroquinoline ≥95%
4-Chloro-3-nitroquinoline

Hazard: Irritant

Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD07369246
CAS Number: 39061-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR