4-Chloro-2-nitropyridine ≥95%

Supplier: Matrix Scientific

072161-250MG 072161-1G 072161-5G
102523-416EA 200 USD
102523-416 102523-418 102523-420
4-Chloro-2-nitropyridine ≥95%
4-Chloro-2-nitropyridine

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD04114167
CAS Number: 65370-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR