4-Bromo-3-hydroxypyridine ≥95%
Supplier: Matrix Scientific
072160-1G
072160-5G
102523-412EA
96.92
USD
102523-412
102523-414
4-Bromo-3-hydroxypyridine ≥95%
4-Bromo-3-hydroxypyridine
Hazard: Irritant
Formula:
C₅H₄BrNO MW: 173.9954 g/mol Melting Pt: 134-136°C |
MDL Number:
MFCD03093021 CAS Number: 161417-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...