4-Bromonicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3-cyanopyridine

072159-250MG 072159-1G 072159-5G
102523-406EA 287.69 USD
102523-406 102523-408 102523-410
4-Bromonicotinonitrile ≥95%
4-Bromonicotinonitrile

Hazard: Irritant

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD08686971
CAS Number: 154237-70-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR